Please login to your online account to display your discounted pricing
BrandTech™ VACUUBRAND™ XS Series Rotary Vane Vacuum Pumps
Designed for very low noise and vibration
$4,520.11 - $6,073.71
Specifications
Voltage 120V
Plug Type US
View More Specs
Products
Catalog Number Mfr. No. For Use With (Application) Type Price Quantity    
13-889-136
brandtech™
698136
Freeze dryers up to 6L, benchtop centrifugal concentrators or other low-temperature evaporations of high boiling point solvents, molecular distillations, schlenk lines and other applications where diaphragm pumps cannot provide sufficient vacuum RZ6 Rotary Vane Vacuum Pump Each for $6,073.71
13-889-135
brandtech™
698126
Small freeze dryers, small centrifugal concentrators or other low-temperature evaporations of high boiling point solvents, molecular distillations, and other applications where diaphragm pumps cannot provide sufficient vacuum RZ2.5 Rotary Vane Vacuum Pump Each for $4,520.11
Description & Specifications
Specifications
Voltage 120V
Plug Type US
  • Small and energy efficient: Substantially smaller and lighter than pumps of comparable capacity, saving you bench or floor space. Motors were selected for their energy efficiency.
  • Rugged and low maintenance: Features many nitrogen-plasma treated parts in flowpath to increase corrosion resistance. Integrated gas ballast helps ensure exceptional water vapor tolerance with minimal impact on ultimate vacuum. Wearing forces have also been reduced for longer service interval