BrandTech™ VACUUBRAND™ MD1C + AK + EK Oil-Free Chemistry Diaphragm Vacuum Pump
An excellent choice for rotary evaporation and other laboratory applications
Manufacturer:  BrandTech™ 20696633
 View more versions of this product
Catalog No. 13-688-286
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Suitable for benchtop rotary evaporators, and powerful enough for excellent results with high-boiling point solvents
  • Extremely low maintenance, no oil changes, no oil mist
  • Ultimate vacuum 1.5 Torr; free air capacity 25 liters per minute
  • Corrosion resistance
  • With an integrated gas ballast to provide consistent pump performance with condensed vapors
  • Adds solvent recovery, including an emissions condenser; compact three-stage design is suitable for benchtop rotary evaporation and gel drying with an SDS-PAGE concentration greater than 10%
Order Info
Included solvent recovery glassware for use in evaporative applications when no cold trap is used.
Specifications
Specifications
16 psi
0.01 hp
Manual
CAN/CSA C22.2 61010-1-12, CE, DIN EN 1012-2:2011, DIN EN 12100:2001, DIN EN 50581:2013, DIN EN 61010-1:2011, DIN EN 61326-1:2013, IEC 61010-1:2010, NRTL Certification, TUV, UL 61010A 61010-1:2012
12.62 x 11.4 x 19 in. (32 x 29 x 48cm)
MD1C+AK+EK
22.48 lb.
104°F
60Hz
175.26 mm
32cm
142.24 mm
IP40
Chemical-Resistant
Rotary Evaporation, Rotary Evaporators, Vacuum Drying Ovens
3
1.5 (without Gas Ballast) torr, 3 (with Gas Ballast) torr
120 V
150 to 1800 RPM
120V
PTFE (Pump Head), Carbon Reinforced PTFE (Case)
45 dB(A)
10.2kg
0.77/0.88 CFM
19 in.
12.44 in.
11.4 in.
Diaphragm Pump
4
50°F
Oil-Free