Please login to your online account to display your discounted pricing
BrandTech™ VACUUBRAND™ Absolute Pressure Vacuum Gauges
Replace fragile McLeod and dial gauges
$1,635.85 - $3,446.09
Specifications
Product Type Absolute Pressure Vacuum Gauge
View More Specs
Includes:  Gauge, mounting rod, operating manual
Products
Catalog Number Mfr. No. Dimensions (L x W x H) Includes Pressure Range Price Quantity    
13-878-81
brandtech™
682902
2.5 x 4.5 x 4.5 in. (6.3 x 11.5 x 11.5cm) 9V Lithium battery with adjustable sleep timer, gauge, mounting rod, operating manual Upper measuring limit:1080mbar / 810 torr; Lower measuring limit: 1 mbar / 1 torr Each for $1,635.85
13-878-232
brandtech™
683170
5.4 x 4.8 x 4.5 in. (13.8 x 12.4 x 11.5cm) Gauge, mounting rod, operating manual Upper measuring limit:1080mbar/810 torr; Lower measuring limit: 0.1 mbar/0.1 torr Each for $3,446.09
Description & Specifications
Specifications
Product Type Absolute Pressure Vacuum Gauge
Displays combine digital indication for precision and analog indication for trending.
  • User-selectable units: torr, mbar, or hectoPascal (hPa)
  • All models can be calibrated for ISO 9000/GLP compliance
  • With capacitive transducers for gas-independent measuring
  • Transducers are made of alumina ceramic with outstanding corrosion resistance and superior long-term stability
  • DVR2 operates on included 9V lithium battery with adjustable sleep timer, and is suitable for central vacuum systems, diaphragm vacuum pumps, and aspirators
  • DVR2 dimensions: 2.5L × 4.5W × 4.5 in.H (6.5 × 11.5 × 11.5cm)
  • DCP3000 + VSK3000 plugs into a standard 115V wall outlet, measures to 0.1 torr/mbar/hPa, and has an RS-232 output
DCP3000 units can be fitted with additional gauge heads for monitoring multiple applications, or increasing accuracy across the atmosphere to 10-3 range. Ask your Fisher Scientific™ representative fro orderign information.
One year
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.