BRAND™ Transferpettor™ Replacement Seals
For use with the Transferpettor™ line, specially designed to handle viscous, foaming liquids and high vapor pressure.
Manufacturer:  BRAND™ 702870
 View more versions of this product
Catalog No. 13-688-124
Description
Description
  • Aids in achieving precision and accuracy
  • For the most demanding pipetting operations
  • Transferpettors operates on the positive displacement principle, with piston in direct contact with the aspirated liquid
  • Provided with DE-M marking
  • Supplied in packs of 10
Specifications
Specifications
DE-M
HDPE
For 1000 to 5000µL Transferpettor
1000 to 5000 µL Transferpettor
10/Pk.