Please login to your online account to display your discounted pricing
BrandTech™ Transferpette™ S Single Channel Fixed Volume Pipet
Uses innovative and ergonomic one-handed operation. BrandTech™ Transferpette™ S Single Channel Fixed Volume Pipet offers robust yet lightweight construction. Easy Calibration™ technology provides long lasting reliability, accuracy, and precision.
$336.79 - $336.79
Specifications
Autoclavable Yes
Certifications/Compliance ISO9001, CE, IVD, certificate of performance, individual calibration certificate
Includes Centralized piston button design, shelf mount, 4 digit display, steam sterilizable shaft, operating manual
Warranty 3 Years
View More Specs
Products
Catalog Number Mfr. No. Accuracy Model Volume (Metric) Price Quantity    
03-837-314
brandtech™
704738
±0.6% F100 100μL Each for $336.79
03-837-316
brandtech™
704754
±0.6% F500 500μL Each for $336.79
03-837-762
brandtech™
704762
±0.6% F1000 1000μL Each for $336.79
03-837-312
brandtech™
704720
±0.8% F25 25μL Each for $336.79
03-837-311
brandtech™
704716
±0.8% F20 20μL Each for $336.79
03-837-313
brandtech™
704728
±0.8% F50 50μL Each for $336.79
03-837-315
brandtech™
704744
±0.6% F200 200μL Each for $336.79
03-837-310
brandtech™
704708
±1% F10 10μL Each for $336.79
Description & Specifications
Specifications
Autoclavable Yes
Certifications/Compliance ISO9001, CE, IVD, certificate of performance, individual calibration certificate
Includes Centralized piston button design, shelf mount, 4 digit display, steam sterilizable shaft, operating manual
Warranty 3 Years
Device is lightweight with one-handed volume adjustment.
  • Centralized pipetting button with optimized stroke length
  • Easy in-lab service
  • Robust and lightweight
  • Accepts universal tips
  • Autoclavable at 121°C/250°F (20 minutes)