BrandTech™ Transferpette™ S Single Channel Fixed Volume Pipet
Uses innovative and ergonomic one-handed operation. BrandTech™ Transferpette™ S Single Channel Fixed Volume Pipet offers robust yet lightweight construction. Easy Calibration™ technology provides long lasting reliability, accuracy, and precision.
Manufacturer:  BrandTech™ 704744
 View more versions of this product
Catalog No. 03-837-315
Description
Description
Device is lightweight with one-handed volume adjustment.
  • Centralized pipetting button with optimized stroke length
  • Easy in-lab service
  • Robust and lightweight
  • Accepts universal tips
  • Autoclavable at 121°C/250°F (20 minutes)
Specifications
Specifications
±0.6%
ISO9001, CE, IVD, certificate of performance, individual calibration certificate
2 to 200μL tip, 5 to 300μL tip, 5 to 200μL tip
3 Years
200μL
Yes
2 to 200μL tip, 5 to 300μL tip, 5 to 200μL tip
Centralized piston button design, shelf mount, 4 digit display, steam sterilizable shaft, operating manual
F200
Documents
Accessories