Please login to your online account to display your discounted pricing
BrandTech™ Transferpette™ S Single Channel Adjustable Pipet
Exibits a high degree of accuracy, precision, maximum versatility, and reliable performance with Easy Calibration™ technology. BrandTrade; Transferpette™ S Single Channel Adjustable Pipet offers simple one-handed operation with an adjustable brake to protect against accidental volume changes.
Manufacturer: brandtech™  704768
 View more versions of this product
Catalog No. 03-837-317
  • / Each
Description & Specifications
Specifications
Includes Performance certificate, operating manual, shelf mount, and three year warranty.
Product Type Single channel pipet
Accuracy ±2%
Autoclavable Yes
Certifications/Compliance CE, IVD
Compatible Tips 0.1 to 20μL and 0.1 to 1μL Nano-cap™ tip
Display Digital
Finger Style Curved
For Use With (Application) Demanding laboratory applications
For Use With (Equipment) 0.1 to 20μL and 0.1 to 1μL Nano-cap™ tip
Increments 0.001μL
Warranty 3 Years
Model D-1
Volume (Metric) 0.1 to 1μL
Compatibility 0.1 to 20μL and 0.1 to 1μL Nano-cap™ tip
The Transferpette™ S Single Channel Adjustable Pipet offers robust yet lightweight construction, simple, one-handed operation, a high degree of accuracy and precision with Easy Calibration™ technology for long lasting reliability. The single channel model is available in fixed volume and digital models in volume ranges from 0.1µL to 10mL.
  • Centralized pipetting button with optimized stroke length
  • Adjustable brake to fix volume range
  • Visible four-digit volume display
  • Compatible with universal tips (0.1-1μL requires nano-cap tips)
  • Repeatedly autoclavable at 121°C/250°F(20min.)