Please login to your online account to display your discounted pricing
BrandTech™ Transferpette™ S Single Channel Adjustable Pipet
Exibits a high degree of accuracy, precision, maximum versatility, and reliable performance with Easy Calibration™ technology. BrandTrade; Transferpette™ S Single Channel Adjustable Pipet offers simple one-handed operation with an adjustable brake to protect against accidental volume changes.
$607.66 - $668.21
Specifications
Autoclavable Yes
Certifications/Compliance CE, IVD
Display Digital
Finger Style Curved
For Use With (Application) Demanding laboratory applications
View More Specs
Includes:  Shelf/rack mount, performance certificate, operating manual
Products
Catalog Number Mfr. No. Compatible Tips Includes Increments Volume (Metric) Price Quantity    
03-837-318
brandtech™
704770
0.1 to 20μL Nano-cap™ tip, 0.5 to 20μL ultra micro tip, 0.1 to 1μL Nano-cap™ tip, 0.5 to 10μL tip Performance certificate, operating manual, shelf mount, and three year warranty. 0.01μL 0.5 to 10μL Each for $607.66
03-837-323
brandtech™
704782
0.5 to 5mL tip Performance certificate, operating manual, shelf mount, and three year warranty. 5μL 500 to 5000μL Each for $607.66
03-837-319
brandtech™
704772
2 to 200μL tip, 5 to 300μL tip, 2 to 20μL tip, 5 to 100μL tip Performance certificate, operating manual, shelf mount, and three year warranty. 0.02μL 2 to 20μL Each for $607.66
03-837-322
brandtech™
704780
50 to 1000μL tip, 50 to 1250μL tip, 50 to 1000μL tip Performance certificate, operating manual, shelf mount, and three year warranty. 1μL 100 to 1000μL Each for $607.66
03-837-321
brandtech™
704778
2 to 200μL tip, 5 to 300μL tip, 2 to 20μL tip, 5 to 100μL tip, 5 to 200μL tip Performance certificate, operating manual, shelf mount, and three year warranty. 0.2μL 20 to 200μL Each for $609.40
03-837-325
brandtech™
704784
1 to 10mL tip Performance certificate, operating manual, shelf mount, and three year warranty. 10μL 1000 to 10,000μL Each for $607.66
03-837-320
brandtech™
704774
2 to 200μL tip, 5 to 300μL tip, 2 to 20μL tip, 5 to 100μL tip Performance certificate, operating manual, shelf mount, and three year warranty. 0.1μL 10 to 100μL Each for $607.66
13-689-204
brandtech™
704769
0.1 to 20μL, 0.5 to 20μL, 1 to 50μL, 0.5 - 10μL, and 1 to 20μL Performance certificate, operating manual, shelf mount, and three year warranty. 0.002μL 0.1 to 2.5μL Each for $609.40
03-837-317
brandtech™
704768
0.1 to 20μL and 0.1 to 1μL Nano-cap™ tip Performance certificate, operating manual, shelf mount, and three year warranty. 0.001μL 0.1 to 1μL Each for $668.21
Description & Specifications
Specifications
Autoclavable Yes
Certifications/Compliance CE, IVD
Display Digital
Finger Style Curved
For Use With (Application) Demanding laboratory applications
Includes Performance certificate, operating manual, shelf mount, and three year warranty.
Warranty 3 Years
The Transferpette™ S Single Channel Adjustable Pipet offers robust yet lightweight construction, simple, one-handed operation, a high degree of accuracy and precision with Easy Calibration™ technology for long lasting reliability. The single channel model is available in fixed volume and digital models in volume ranges from 0.1µL to 10mL.
  • Centralized pipetting button with optimized stroke length
  • Adjustable brake to fix volume range
  • Visible four-digit volume display
  • Compatible with universal tips (0.1-1μL requires nano-cap tips)
  • Repeatedly autoclavable at 121°C/250°F(20min.)
Three-year warranty