BrandTech™ Transferpette™ S Adjustable Single Channel Pipet Starter Sets
Simple one-handed operation units exibits high degree accuracy, precision, maximum versatility and reliable performance with Easy Calibration™ technology
$1,481.40 - $1,481.40
Specifications
Autoclavable Yes
For Use With (Application) Demanding laboratory applications
View More Specs
Includes: Set contents:
  • Three adjustable volume pipettes
  • Three shelf/rack mounts
  • Three filled polypropylene Tip Boxes
Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Product Type Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Includes Product Type Volume (Metric) Price Quantity  
03-837-327 Accessories BrandTech™
704791
D-20, D-200, D-1000 adjustable volume pipette, three shelf/rack mounts, three filled Polypropylene tip boxes Midsize Set 2 to 20, 20 to 200, 100 to 1000μL Each for $1,481.40
03-837-328 BrandTech™
704792
D1000, D5000, D10000 adjustable volume pipette, three shelf/rack mounts, three filled Polypropylene tip boxes Macro Set 100 to 1000, 500 to 5000, 1000 to 10000μL Each for $1,481.40
03-837-790 BrandTech™
704790
D-1, D-10, D-100 adjustable volume pipette, three shelf/rack mounts, three filled Polypropylene tip boxes Micro Set 0.1 to 1, 0.5 to 10, 10 to 100μL Each for $1,481.40
03-837-793 BrandTech™
704793
D-10, D-100, D-1000 adjustable volume pipette, three shelf/rack mounts, three filled Polypropylene tip boxes Standard Set 0.5 to 10, 10 to 100, 100 to 1000μL Each for $1,481.40
Specifications
Description
  • Available as Standard Set, Micro Set, Midsize Set and Macro Set
Description & Specifications
Specifications
Yes
Demanding laboratory applications