Please login to your online account to display your discounted pricing
BrandTech™ Transferpette™ S Adjustable Single Channel Pipet Starter Sets
Simple one-handed operation units exibits high degree accuracy, precision, maximum versatility and reliable performance with Easy Calibration™ technology
Manufacturer: brandtech™  704792
 View more versions of this product
Catalog No. 03-837-328
  • / Each
Description & Specifications
Specifications
Product Type Macro Set
Volume (Metric) 100 to 1000, 500 to 5000, 1000 to 10000μL
Includes D1000, D5000, D10000 adjustable volume pipette, three shelf/rack mounts, three filled Polypropylene tip boxes
For Use With (Application) Demanding laboratory applications
Autoclavable Yes