BrandTech™ Transferpette™ S Multichannel Pipet
Protects against accidental volume changes using innovative one-handed volume adjustment with brake. BrandTech™ Transferpette™ S Multichannel Pipet is supplied with performance certificate, operating manual, shelf mount, two tip boxes, reagent reservoir, pack of replacement v-rings, and one year warranty. Patented individually removable/replaceable tip cones provide easy in-lab cleaning.
$1,443.45 - $1,911.36
Specifications
Autoclavable Yes
Dimensions (L x W x H) 241.3 x 142.8 x 123.8mm
Display Digital
Includes Shelf/rack mount, box of pipette tips with two refills, set of replacement V-rings, certificate of performance and comprehensive operating manual
Stroke (Metric) Length 12.5mm
View More Specs
Includes: Shelf/rack mount, box of pipette tips with two refills, set of replacement V-rings, certificate of performance and comprehensive operating manual
Products ${productFamilyLength}
Catalog Number Mfr. No. Accuracy Number of Channels Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Accuracy Number of Channels Volume (Metric) Price Quantity  
03-837-306 BrandTech™
703720
±1.6% 12 0.5 to 10μL Each for $1,781.51
03-837-326 BrandTech™
703700
±1.6% 8 0.5 to 10μL Each for $1,443.45
03-837-329 BrandTech™
703710
±0.8% 8 20 to 200μL Each for $1,443.45
03-837-732 BrandTech™
703732
±0.6% 12 30 to 300μL Each for $1,479.49
03-837-301 BrandTech™
703706
±0.8% 8 5 to 50μL Each for $1,443.45
03-837-308 BrandTech™
703728
±0.8% 12 10 to 100μL Each for $1,781.51
03-837-307 BrandTech™
703726
±0.8% 12 5 to 50μL Each for $1,911.36
03-837-309 BrandTech™
703730
±0.8% 12 20 to 200μL Each for $1,781.51
03-837-305 BrandTech™
703712
±0.6% 8 30 to 300μL Each for $1,443.45
03-837-302 BrandTech™
703708
±0.8% 8 10 to 100μL Each for $1,443.45
Specifications
Description
Device has one-handed volume adjustment with brake for secure setting.
  • With central button control layout
  • Short, 12.5mm piston stroke-length
  • V-rings for low-force tip mounting and ejection
  • Four digit volume display
  • Lightweight
  • Individually removable/replaceable patented tip cones
  • Autoclavable at 121°C (20 minutes)
Description & Specifications
Specifications
Yes
Digital
12.5mm
241.3 x 142.8 x 123.8mm
Shelf/rack mount, box of pipette tips with two refills, set of replacement V-rings, certificate of performance and comprehensive operating manual
1 Year