missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BrandTech™ Transferpette™ S Multichannel Pipet

Protects against accidental volume changes using innovative one-handed volume adjustment with brake. BrandTech™ Transferpette™ S Multichannel Pipet is supplied with performance certificate, operating manual, shelf mount, two tip boxes, reagent reservoir, pack of replacement v-rings, and one year warranty. Patented individually removable/replaceable tip cones provide easy in-lab cleaning.

Manufacturer:  BrandTech™ 703728

 View more versions of this product

Catalog No. 03-837-308


 

Specifications

Description

Pipet, 12 channel; BrandTech; Transferpette S; Lightweight; Central button control layout; 12.5mm piston stroke; w/brake; V-rings; 4 digit volume display; Replaceable tip cones; Easy Calibration; Autoclavable: 121deg.C; 10 to 100uL

Device has one-handed volume adjustment with brake for secure setting.

 

  • With central button control layout
  • Short, 12.5mm piston stroke-length
  • V-rings for low-force tip mounting and ejection
  • Four digit volume display
  • Lightweight
  • Individually removable/replaceable patented tip cones
  • Autoclavable at 121°C (20 minutes)
Description & Specifications

Specifications

±0.8%
241.3 x 142.8 x 123.8mm
4.87 in.
Shelf/rack mount, box of pipette tips with two refills, set of replacement V-rings, certificate of performance and comprehensive operating manual
241.3mm
1 Year
142.8mm
10 to 100μL
Yes
Digital
123.8mm
9.5 in.
12.5mm
5.62 in.
12