Please login to your online account to display your discounted pricing

BrandTech™ Transferpette™ S Multichannel Pipet


Protects against accidental volume changes using innovative one-handed volume adjustment with brake. BrandTech™ Transferpette™ S Multichannel Pipet is supplied with performance certificate, operating manual, shelf mount, two tip boxes, reagent reservoir, pack of replacement v-rings, and one year warranty. Patented individually removable/replaceable tip cones provide easy in-lab cleaning.

Manufacturer: brandtech™  703726

 View more versions of this product

Catalog No. 03-837-307

  • / Each
 

Description & Specifications

Specifications

Dimensions (L x W x H) 241.3 x 142.8 x 123.8mm
Warranty 1 Year
Display Digital
Length (English) 9.5 in.
Height (Metric) 123.8mm
Width (English) 5.62 in.
Number of Channels 12
Stroke (Metric) Length 12.5mm
Volume (Metric) 5 to 50μL
Autoclavable Yes
Length (Metric) 241.3mm
Includes Shelf/rack mount, box of pipette tips with two refills, set of replacement V-rings, certificate of performance and comprehensive operating manual
Height (English) 4.87 in.
Width (Metric) 142.8mm
Accuracy ±0.8%

Device has one-handed volume adjustment with brake for secure setting.

 

  • With central button control layout
  • Short, 12.5mm piston stroke-length
  • V-rings for low-force tip mounting and ejection
  • Four digit volume display
  • Lightweight
  • Individually removable/replaceable patented tip cones
  • Autoclavable at 121°C (20 minutes)