missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BrandTech™ Transferpette™ -8/-12 Electronic Multichannel Pipette
Versatile pipette offering five operating modes to improve productivity and increase sample throughput.
Supplier: BrandTech™ 705419
Description
- Choice of 8 or 12 channel models
- Clear, legible LCD screen
- Intuitive interface with user-friendly design allows easy operation
- Soft touch pipetting key and adjustable finger rest provides added comfort
- Rechargeable battery provides over 4000 pipetting cycles between charges
- Multichannel V-rings on tip cones improves tip fit and reduces mounting and ejection force
- Easily maintained
Operating modes
- Pipetting (PIP Mode): previously set volume is aspirated into pipette tip and then discharged
- Mixing of samples (PIPmix Mode): program for mixing liquids
- Reverse Pipetting (revPIP Mode): program designed for greater accuracy
- Pipetting for electrophoresis (GEL Mode): program for loading of electrophoresis gels
- Dispensing (DISP Mode): single aspirated volume is dispensed in selected number of smaller, equal aliquots
Specifications
±0.12 μL | |
0.1 to 20 μL Nano-cap Tip, 0.5 to 20 μL Ultra Micro Tip, 0.1 to 1 μL Nano-cap Tip, 0.5 to 10 μL Tip | |
8 | |
Step-based | |
0.5 to 10 μL | |
Ergonomic | |
0.02 μL | |
1 Year |
Rechargeable | |
DE-M | |
Transferpette | |
Electronic Pipette | |
LCD | |
Battery, Power Supply Unit, 120 VAC Adapter with Charging Cable, Operating Manual, Pipette Stand, Reagent Reservoir | |
Yes |