missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BrandTech™ Transferpette™ -8/-12 electronic Multichannel Pipette

Versatile pipette offers five operating modes to improve productivity and increase sample throughput

Manufacturer:  BrandTech™ 705424

 View more versions of this product

Catalog No. 13-668-428



Includes: Battery, 120V AC adapter with charging cable, silicone oil, operating manual, pipette stand, one Tip-Box, one TipRefill, reagent reservoir, tip cone tool, set of replacement V-rings

Description

Description

  • Choice of 8 or 12 channel models
  • Clear, legible LCD screen
  • Intuitive interface with user-friendly desing allows easy operation
  • Soft touch pipetting key and adjustable finger rest provides added comfort
  • Rechargeable battery provides over 4000 pipetting cycles between charges
  • Multichannel V-rings on tip cones improves tip fit and reduces mounting and ejection force
  • Easily maintained

Operating modes

  • Pipetting (PIP Mode): previously set volume is aspirated into pipette tip and then discharged
  • Mixing of samples (PIPmix Mode): program for mixing liquids
  • Reverse Pipetting (revPIP Mode): program designed for greater accuracy
  • Pipetting for electrophoresis (GEL Mode): program for loading of electrophoresis gels
  • Dispensing (DISP Mode): single aspirated volume is dispensed in selected number of smaller, equal aliquots

Warranty

One-year

Specifications

Specifications

±1.6μL
2 to 200μL tip, 5 to 300μL tip, 2 to 20μL tip, 5 to 100μL tip, 5 to 200μL tip
Ergonomic
0.2μL
Step Based
Transferpette -8/-12 electronic
10 to 200μL
Rechargeable
LCD
Battery, power supply unit, 120VAC adapter with charging cable, one pipette tip sample, operating manual, pipette stand, tip-box SL filled with appropriate tip, two tip refill inserts, reagent reservoir and five operating modes
Yes
1 Year
8
10 to 200μL
Documents