missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BrandTech™ Support Jacks
Available in anodized aluminum (blue) or epoxy-powder-coated aluminum (green) decks
Supplier: BrandTech™ B11080
Description
- Lab Support Jack has a 30 x 30 cm deck made of 304 stainless steel
- Lift mechanism is made of 304 stainless steel with POM rollers for long service life Minimum height 9 cm, maximum height maximum height 47 cm.
- Maximum static load, 60 kg.
Specifications
Epoxy-powder Coated Aluminum | |
3.5 to 18.5 in. | |
30 cm | |
30 cm | |
Support Jack |
9 to 47 cm | |
11.8 in. | |
11.8 in. | |
132.27 lb. | |
Fume hoods and marine and industrial laboratories, POM rollers |