BrandTech™ Telescoping Filling Tube for Dispensette™III, Titrette™, and seripettor™ pro
Manufactured from FEP. BrandTech™ Telescoping Filling Tube for Dispensette™III, Titrette™, seripettor™ pro adjusts easily to different size bottles without measuring or cutting. Available in a variety of lengths and fits a wide range of bottles.
$63.54 - $116.51
Specifications
Material FEP
Product Type Telescopic Filling Tube
For Use With (Equipment) Dispensette™ III, Titrette™, seripettor™ pro
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity (Metric) Diameter (Metric) Outer Neck Length (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Diameter (Metric) Outer Neck Length (Metric) Price Quantity  
13-879-77 BrandTech™
704201
0.5, 1, 2, 5, 10mL 6mm 250 to 480mm Each for $80.60
13-879-79 BrandTech™
704203
0.5, 1, 2, 5, 10mL 6mm 125 to 240mm Each for $63.54
13-880-709 BrandTech™
704208
0.5, 1, 2, 5, 10mL 6mm 195 to 350mm Each for $116.51
Specifications
Description
  • Manufactured from FEP
  • Adjusts easily to different size bottles without measuring or cutting.
  • For use with Dispensette™III, Titrette™, and seripettor™ pro.
  • Available in a variety of lengths
  • Fits a wide range of bottles.
Description & Specifications
Specifications
FEP
Telescopic Filling Tube
Dispensette™ III, Titrette™, seripettor™ pro