missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BrandTech™ Plastic Cuvets
Excellent choices for most spectrophotometry applications in the visible range.
Manufacturer: BrandTech™ 759075DE
Description
- Easy to use with an arrow mark to indicate direction of transmission
- Range of application: 340nm or longer for polystyrene, 300nm or longer for methacrylate; 10mm lightpath
- Macro and micro sizes available (1.5 - 4.5mL)
- Mold cavity matched for lowest variation in extinction coefficient
Specifications
Spectrophotometer Cuvette | |
3 mL | |
500 Pack | |
340nm | |
For most spectrophotometry applications in the visible range, easy to use with an arrow mark to indicate direction of transmission |
Yes | |
Polystyrene | |
10 mm | |
Most spectrophotometers and photometers |