BrandTech™ Flexible Discharge Tubes for Dispensette™ III Bottle-Top Dispensers
Enhances the suitability of Dispensette dispensers for various applications.
Manufacturer:  BrandTech™ 707925
 View more versions of this product
Catalog No. 13-688-77
Description
Description
  • For serial dispensing such as scintillation cocktail, the flexible discharge tube preserves all important safety innovations
  • PTFE material
  • Fine tip
Order Info
Not for use with the Dispensette HF or dispensers from other manufacturers.
Specifications
Specifications
Flexible Discharge Tube
Serial dispensing such as scintillation cocktail, the flexible discharge tube preserves all important safety innovations
31.5 in.
800mm
1 to 10mL Dispensette™ III, Dispensette™ organic bottletop dispenser
Safety handle
PTFE
Fine Tip