Please login to your online account to display your discounted pricing
BrandTech™ Erlenmeyer Flasks with Screw Caps
$165.00 - $198.36
Specifications
Autoclavable Yes
Graduation Marks Printed scale
Mouth Style Wide
View More Specs
Products
Catalog Number Mfr. No. Graduation (Metric) Capacity (Metric) Material Price Quantity    
14-379-191
brandtech™
V670941
10mL 1000mL Polypropylene Pack of 4 Each for $198.36
14-379-195
brandtech™
V668941
50mL 250mL Polypropylene Pack of 6 Each for $165.00
Description & Specifications
Specifications
Autoclavable Yes
Graduation Marks Printed scale
Mouth Style Wide