BrandTech™ Class A Volumetric PFA Flasks with Screw Caps
VITLAB PFA volumetric flasks are individually calibrated, and certified Class A to DIN EN ISO 1042.
$1644.91 - $2923.71
Specifications
Calibration Calibrated To Contain
Closure Material Perfluoxy Copolymer
Color Highly Transparent
Description Individually calibrated ring-marking
Material PFA
View More Specs
Products 3
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
14-379-206
View Documents Promotion Details
BrandTech™
107297
50 mL
Pack of 2 Each for $1,644.91
14-379-205
View Documents Promotion Details
BrandTech™
107597
500 mL
Pack of 2 Each for $2,923.71
14-379-367
View Documents Promotion Details
BrandTech™
107097
10 mL
Each for $1,691.34
Description
Description
  • Highly translucent with highly legible ring marking to assist meniscus adjustment
  • PFA screw caps help protect against outside contamination and leakage
Specifications
Specifications
Calibrated To Contain
Highly Transparent
PFA
-200°C to +260°C
Perfluoxy Copolymer
Individually calibrated ring-marking
-328°F to +500°F
Autoclavable
SDS
missing translation for 'documents'
missing translation for 'documents'