Please login to your online account to display your discounted pricing
BrandTech™ BRANDplates™ pureGrade™ S 1536-well Microplates
Non-treated sterile microplates for diverse applications
$3,797.82 - $3,797.82
Specifications
Sterile Yes
Volume (Metric) Well 10μL
Bottom Flat (F)
View More Specs
Products
Catalog Number Mfr. No. Color Price Quantity    
14-386-006
brandtech™
781700
Transparent Pack of 50 Each for $3,797.82
14-386-007
brandtech™
781701
White Pack of 50 Each for $3,797.82
14-386-008
brandtech™
781702
Black Pack of 50 Each for $3,797.82
Description & Specifications
Specifications
Sterile Yes
Volume (Metric) Well 10μL
Bottom Flat (F)
  • Nontreated PS surface
  • Medium binding
  • Sterilized via beta-radiation
  • Sterile plate for most applications
  • With lid
  • Available in clear, white or black
  • Free from endotoxins, DNase, DNA, RNase; noncytotoxic
Bacteriological assays, screenings, storage.