Please login to your online account to display your discounted pricing
BrandTech™ BRANDplates™ lipoGrade™ 384-Well Microplates
Immunoassay plate optimized for the immobilization of hydrophobic molecules
$639.32 - $1,799.96
Specifications
Sterile No
Well Design Flat (F)
View More Specs
Products
Catalog Number Mfr. No. Color Volume (Metric) Well Price Quantity    
14-386-037
brandtech™
781861
White 0.1mL Pack of 50 Each for $858.86
14-386-036
brandtech™
781860
Transparent 0.1mL Pack of 50 Each for $639.32
14-386-038
brandtech™
781862
Black 0.1mL Pack of 50 Each for $858.86
14-386-042
brandtech™
781866
White 0.12mL Pack of 50 Each for $1,799.96
14-386-043
brandtech™
781867
Black 0.12mL Pack of 50 Each for $1,799.96
Description & Specifications
Specifications
Sterile No
Well Design Flat (F)
  • High binding for molecules with predominately hydrophobic regions such as lipoproteins or peptides
  • PS, non-sterile
  • For solid phase assays with hydrophobic molecules and liquid phase with hydrophilic molecules, respectively
  • Alternative to immunoGrade™ surface
  • Available in transparent, white or black color
  • With standard or transparent bottom
  • Modification via physical-chemical process, good shelf life at room temperature
  • Free from endotoxins, DNase, DNA, RNase, non-cytotoxic