Please login to your online account to display your discounted pricing
BrandTech™ BRANDplates™ immunoGrade™ 384-Well Microplates
Immunoassay plate optimized for immobilization of IgG
$674.82 - $674.82
Specifications
Sterile No
Well Design Flat (F)
Volume (Metric) Well 100μL
View More Specs
Products
Catalog Number Mfr. No. Color Price Quantity    
14-386-031
brandtech™
781741
White Pack of 50 Each for $674.82
14-386-030
brandtech™
781740
Transparent Pack of 50 Each for $674.82
Description & Specifications
Specifications
Sterile No
Well Design Flat (F)
Volume (Metric) Well 100μL
  • High binding for molecules with mixed hydrophilic and hydrophobic regions
  • Surface choice for majority of standard ELISAs
  • For solid phase immunoassays
  • PS, non-sterile
  • Available in clear, white or black
  • Free from endotoxins, DNase, DNA, RNase, non-cytotoxic