BRAND™ BRANDplates™ immunoGrade™ 384-Well Microplates
Immunoassay plate optimized for immobilization of IgG
Description
- High binding for molecules with mixed hydrophilic and hydrophobic regions
- Surface choice for majority of standard ELISAs
- For solid phase immunoassays
- PS, non-sterile
- Available in clear, white or black
- Free from endotoxins, DNase, DNA, RNase, non-cytotoxic
Specifications
Standard | |
16 x 24 | |
Polystyrene | |
Non-sterile | |
For immobilization of IgG, solid phase immunoassays |
Without Lid | |
Clear | |
384 | |
Flat Bottom (F-well) | |
50/Cs. |