Please login to your online account to display your discounted pricing
BrandTech™ BRANDplates™ hydroGrade™ 384-Well Microplates
Immunoassay plate optimized for immobilization of hydrophilic molecules
$824.61 - $1,006.07
Specifications
Sterile No
Well Design Flat (F)
Volume (Metric) Well 0.1mL
View More Specs
Products
Catalog Number Mfr. No. Color Price Quantity    
14-386-035
brandtech™
781802
Black Pack of 50 Each for $1,006.07
14-386-034
brandtech™
781801
White Pack of 50 Each for $1,006.07
14-386-033
brandtech™
781800
Transparent Pack of 50 Each for $824.61
Description & Specifications
Specifications
Sterile No
Well Design Flat (F)
Volume (Metric) Well 0.1mL
  • High affinity for molecules with hydrophilic regions such as glycoproteins, peptides and more
  • PS, nonsterile
  • For solid phase assays with hydrophilic molecules and liquid phase with hydrophobic molecules, respectively
  • Alternative to immunoGrade™ surface
  • Available in clear, white or black
  • Free from endotoxins, DNase, DNA, RNase, noncytotoxic