Please login to your online account to display your discounted pricing
BrandTech™ BRANDplates™ cellGrade™ premium 384-Well Microplates
For serum-free and serum-reduced cultivation of cells
$2,221.79 - $3,650.11
Specifications
Sterile Yes
Well Design Flat (F)
View More Specs
Products
Catalog Number Mfr. No. Color Volume (Metric) Well Price Quantity    
14-386-055
brandtech™
782100
Transparent 100μL Pack of 50 Each for $2,221.79
14-386-056
brandtech™
782106
White 120μL Pack of 50 Each for $3,650.11
14-386-057
brandtech™
782107
Black 120μL Pack of 50 Each for $3,650.11
Description & Specifications
Specifications
Sterile Yes
Well Design Flat (F)
  • Poly-D-Lysine-equivalent surface with analogous results regarding cell growth and morphology
  • Optimized cell adhesion reduces cell damage from frequent washing
  • Alternative option to biologically coated surfaces
  • Physical/chemical treatment has long room temperature shelf life compared to biological coatings
  • Available in clear, white or black with standard or transparent F-bottom and lid
  • Sterilized by beta-radiation
  • DNA-, DNase-, RNase-free; noncytotoxic
  • Endotoxins are <0.01EU/mL
Meet requirements of Ph. Eur. and USP 29.