Please login to your online account to display your discounted pricing

BrandTech™ BRAND™ Pipet Tips, 50 to 1000uL: Ultra-Low Retention

Ultra-low retention tips for routine laboratory and research pipetting

$115.60 - $206.63

Specifications

Certifications/Compliance DNA, RNase, ATP and endotoxin-free
Color Colorless
Material Polypropylene
Sterility Nonsterile
For Use With (Application) For routine laboratory and research pipetting
View More Specs
Products
Catalog Number Mfr. No. Graduations Volume (Metric) Price Quantity    

13-889-195

 
brandtech™
732312
2, 10μL 0.5 to 10μL Pack of 480 Each for $115.60

13-889-217

 
brandtech™
732352
250, 500, 1000μL 50 to 1000μL Pack of 960 Each for $206.63
Description & Specifications

Specifications

Certifications/Compliance DNA, RNase, ATP and endotoxin-free
Color Colorless
Material Polypropylene
Sterility Nonsterile
For Use With (Application) For routine laboratory and research pipetting

  • High-purity polypropylene is oleamide- and diHEMDA-free
  • Nonsterile
  • All unopened packages are DNA-, RNase-, ATP- and endotoxin-free
  • Thin-walled for high translucency, easy ejection and low waste
  • Graduations at 250, 500 and 1000μL for quick volume check
  • In convenient, ecological rack-and-stack format
  • Tips are colorless and placed into blue-colored tip tray

  • Produced through special, patented physicochemical process
  • Homogeneous, defect-free surface for low surface tension and low retention
  • Reduce sample loss and provide substantially higher reproducibility when working with critical media
  • For biological samples that contain proteins, nucleic acids, detergents or other difficult liquids that can increase sample loss
  • High chemical resistance
  • Tips can be autoclaved at 121°C (2 bar) without damaging low retention properties