BrandTech™ BRAND™ Pipet Tips, 2 to 200uL: Ultra-Low Retention
Low retention tips used for virtually all pipettes with yellow color code
$71.34 - $125.43
Specifications
Certifications/Compliance DNA, RNase, ATP and endotoxin-free
Color Colorless
Material Polypropylene
Sterility Nonsterile
Product Type Tip
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Graduations Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Graduations Volume (Metric) Price Quantity  
13-889-193 BrandTech™
732308
2, 10μL 0.5 to 10μL Pack of 480 Each for $71.34
13-889-216 BrandTech™
732348
20, 100μL 2 to 200μL Pack of 960 Each for $125.43
Specifications
Description
  • High-purity polypropylene is oleamide- and diHEMDA-free
  • Nonsterile
  • All unopened packages are DNA-, RNase-, ATP- and endotoxin-free
  • Thin-walled for high translucency, easy ejection and low waste
  • Graduations at 20 and 100μL for quick volume check
  • Available in ecological rack-and-stack formats
  • Colorless and placed into yellow-colored tip tray
  • Produced through special, patented physicochemical process
  • Homogeneous, defect-free surface thus produced has low surface tension
  • Reduce sample loss and provide substantially higher reproducibility when working with critical media
  • For biological samples that contain detergents such as Triton™ X-100, SDS, Tween, etc.
  • High chemical resistance
  • Tips can be autoclaved at 121°C (2 bar) without damaging material properties
Description & Specifications
Specifications
DNA, RNase, ATP and endotoxin-free
Polypropylene
Tip
Colorless
Nonsterile