Please login to your online account to display your discounted pricing

BrandTech™ BRAND™ Pipet Tips, 1 to 50uL

Fits most pipettes that use standard ultra-micro/crystal tips

$77.00 - $715.03

Specifications

Certifications/Compliance DNA, RNase, ATP and endotoxin-free
Color Colorless
Material Polypropylene
Product Type Tip
Sterility Nonsterile
View More Specs
Products
Catalog Number Mfr. No. Product Type Packaging Quantity Price Quantity    

13-889-142

 
brandtech™
732006
Tip Bulk 2000 Pack Pack of 2000 Each for $168.54

13-889-148

 
brandtech™
732026
Tip Bulk 10000 Pack Pack of 10000 Each for $715.03

13-889-154

 
brandtech™
732106
Tip Rack 960 Pack Pack of 960 Each for $127.60

13-889-173

 
brandtech™
732206
Tip Rack 480 Pack Pack of 480 Each for $77.00

13-889-192

 
brandtech™
732306
Tip Rack 480 Pack Pack of 480 Each for $93.91
Description & Specifications

Specifications

Certifications/Compliance DNA, RNase, ATP and endotoxin-free
Color Colorless
Material Polypropylene
Product Type Tip
Sterility Nonsterile
Volume (Metric) 1 to 50μL
For Use With (Application) High translucency, easy ejection and low waste, quick volume check

  • High-purity polypropylene is oleamide- and diHEMDA-free
  • Nonsterile
  • Available in either standard or ultra low retention types
  • Available either in bulk packages or in convenient rack format
  • All unopened packages are DNA-, RNase-, ATP- and endotoxin-free
  • For reverse pipetting of small volumes

Ultra low retention tips

  • Produced through special, patented physicochemical process
  • Homogeneous, defect-free surface with low surface tension for minimal retention
  • Reduce sample loss and provide substantially higher reproducibility when working with critical media
  • For biological samples that contain proteins, nucleic acids, detergents or other difficult liquids where sample loss is possible
  • High chemical resistance
  • Tips can be autoclaved at 121°C (2 bar) without damaging low retention properties