missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BrandTech™ BRANDplates lipoGrade™ 96-Well Microplates

Featuring a strongly hydrophobic/lipophilic surfacehomogeneous assays with hydrophilic molecules that remain in solution

$1,114.68 - $1,114.68

Specifications

Certifications/Compliance ANSI, SLAS
Sterility Non-sterile
Format Standard
For Use With (Application) Immobilization of hydrophobic molecules
Lid Without Lid
View More Specs

Products 1
Catalog Number Mfr. No. Color Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Color Volume (Metric) Price Quantity  
13-882-241 View Documents BrandTech™
BRT781849
Black 350 μL Pack of 100 Each for $1,114.68
 
Description

Description

  • Immunoassay surface for the immobilization of biomolecules with strongly hydrophobic regions
  • Designed to minimize cross-contamination
  • Blue alphanumeric embossing for easy well identification
  • Non-sterile

Order Info

Packaging is 20 stacks of 5 plates each (100 plates). Each stack comes with one lid.

Specifications

Specifications

ANSI, SLAS
Standard
Without Lid
96-Well Microplate
Non-sterile
Immobilization of hydrophobic molecules
Polystyrene
96