missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BrandTech™ BRANDplates cellGrade™ premium 96-Well Microplates

Plates with surface characteristics for the most demanding cell lines

$1,204.36 - $2,468.71

Specifications

Certifications/Compliance ISO 11137, USP 29
Sterile Yes
Surface Coating Poly-D-Lysine Equivalent
Format 96-well
For Use With (Application) Cultivation of cell lines with the highest demands on their environment
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Color Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Color Volume (Metric) Price Quantity  
13-882-257 BrandTech™
782082
Transparent 350μL Pack of 50 Each for $1,204.36
13-882-259 BrandTech™
782088
Black 350μL Pack of 50 Each for $1,933.29
13-882-260 BrandTech™
782090
White 330μL Pack of 50 Each for $2,468.71
13-882-258 BrandTech™
782085
White 350μL Pack of 50 Each for $1,933.29
13-882-261 BrandTech™
782091
Black 330μL Pack of 50 Each for $2,468.71
Specifications

Description

  • Poly-D-Lysine-equivalent surface with analogous results regarding cell growth and morphology
  • Optimized cell adhesion reduces cell damage from frequent washing
  • Allows for serum-free and serum-reduced cultivation
  • Physical/chemical treatment has long room temperature shelf life compared to biological coatings
  • Sterilized by β-radiation to ISO 11137 and AAMI guidelines for a SAL of 10-6
  • Meet requirements of Ph. Eur. and USP 29
  • DNA-, DNase-, RNase-free
  • Endotoxins are <0.01 EU/mL
  • Orange alphanumeric embossing for easy well identification

Order Info

Packs of 50 individually wrapped plates with lids

Description & Specifications

Specifications

ISO 11137, USP 29
Poly-D-Lysine Equivalent
Cultivation of cell lines with the highest demands on their environment
Polystyrene
96
Yes
96-well
Yes
Premium 96-Well Microplate