Please login to your online account to display your discounted pricing

BrandTech™ BRANDplates cellGrade™ plus 96-Well Microplates

Designed for the cultivation of cells in reduced-serum media and fastidious cell lines

$778.46 - $1,517.36

Specifications

Certifications/Compliance ISO 11137, DIN EN ISO 10993
Sterile Yes
Surface Coating Hydrophilic
Format 96-well
For Use With (Application) Cultivation of cells in reduced serum media and fastidious cell lines
View More Specs
Products
Catalog Number Mfr. No. Color Volume (Metric) Price Quantity    

13-882-252

 
brandtech™
782022
Transparent 350μL Pack of 50 Each for $778.46

13-882-254

 
brandtech™
782028
Black 350μL Pack of 50 Each for $1,253.46

13-882-253

 
brandtech™
782025
White 350μL Pack of 50 Each for $1,253.46

13-882-255

 
brandtech™
782030
White 330μL Pack of 50 Each for $1,517.36

13-882-256

 
brandtech™
782031
Black 330μL Pack of 50 Each for $1,517.36
Description & Specifications

Specifications

Certifications/Compliance ISO 11137, DIN EN ISO 10993
Sterile Yes
Surface Coating Hydrophilic
Format 96-well
For Use With (Application) Cultivation of cells in reduced serum media and fastidious cell lines
Lid Yes
Material Polystyrene
Type Plus 96-Well Microplate
No. of Wells 96

  • Hydrophilic surface with freely-accessible amino, carboxyl and hydroxyl groups
  • Protein-like composition allows for direct cellular attachment
  • Sterilized by β-radiation to ISO 11137 and AAMI guidelines for a SAL of 10-6
  • Meet the requirements of Ph. Eur. and the USP 29
  • DNA-, DNase-, RNase-free
  • Endotoxins are <0.01 EU/mL
  • Orange alphanumeric embossing for easy well identification

Packs of 50 individually wrapped plates with lids