missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BrandTech™ BRANDplates cellGrade™ 96-Well Microplates

Designed for the cultivation of adherent cell lines

$485.14 - $508.96

Specifications

Certifications/Compliance ISO 11137, DIN EN ISO 10993
Sterile Yes
Surface Coating Carboxyl and Hydroxyl Groups
Format 96-well
For Use With (Application) Cultivation of adherent cell lines
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Color Volume (Metric) Well Design Price Quantity  
Catalog Number Mfr. No. Color Volume (Metric) Well Design Price Quantity  
13-882-246 BrandTech™
781962
Transparent 350μL Flat (F) Pack of 50 Each for $485.14
13-882-244 BrandTech™
781960
Transparent 330μL U-Bottom Pack of 50 Each for $485.14
13-882-248 BrandTech™
781965
White 350μL Flat (F) Pack of 50 Each for $508.96
13-882-249 BrandTech™
781968
Black 350μL Flat (F) Pack of 50 Each for $500.75
Specifications

Description

  • Hydrophilic surface with freely-accessible carboxyl and hydroxyl groups
  • Easily binds serum components to for indirect adhesion of cells
  • Sterilized by β-radiation to ISO 11137 and AAMI guidelines for a SAL of 10-6
  • Meet the requirements of Ph. Eur. and the USP 29
  • DNA-, DNase-, RNase-free
  • Endotoxins are <0.01 EU/mL
  • Orange alphanumeric embossing for easy well identification

Order Info

Packs of 50 individually wrapped plates with lids

Description & Specifications

Specifications

ISO 11137, DIN EN ISO 10993
Carboxyl and Hydroxyl Groups
Cultivation of adherent cell lines
Polystyrene
96
Yes
96-well
Yes
96-Well Microplate