Please login to your online account to display your discounted pricing
BrandTech™ BRANDplates cellGrade™ 1536-Well Microplates
$3,168.50 - $3,323.43
Specifications
Well Design Flat (F)
Material Polystyrene
View More Specs
Products
Catalog Number Mfr. No. Color Price Quantity    
14-386-009
brandtech™
782000
Transparent Pack of 50 Each for $3,168.50
14-386-010
brandtech™
782001
White Pack of 50 Each for $3,323.43
14-386-011
brandtech™
782002
Black Pack of 50 Each for $3,323.43
Description & Specifications
Specifications
Well Design Flat (F)
Material Polystyrene
  • Hydrophilic surface with freely-accessible carboxyl and hydroxyl groups
  • Easily binds serum components to for indirect adhesion of cells
  • Sterilized by β-radiation to ISO 11137 and AAMI guidelines for a SAL of 10-6
  • Meet the requirements of Ph. Eur. and the USP 29
  • DNA-, DNase-, RNase-free; noncytotoxic
  • Endotoxins are <0.01 EU/mL
  • For standard cell culture applications
  • Individually wrapped with lid