BrandTech™ BRAND™ Dispensette™ TA Bottletop Dispensers
Provides outstanding performance for precise-volume dispensing of high purity chemicals for trace analysis
Manufacturer:  BrandTech™ 707956
 View more versions of this product
Catalog No. 13-688-336
Includes: Replacement dispenser tips for BRAND™ Dispensette™ TA Bottletop Dispensers
Description
Description
Replacement dispenser tips provide outstanding performance for precise-volume dispensing of high purity chemicals for trace analysis
† Note: Hydrofluoric acid reacts slightly with sapphire. Therefore a minor increase of the aluminum level may occur.
Specifications
Specifications
Discharge Tube
Integrated valve, tantalum valve spring
90mm
Dispensette™ TA bottletop dispenser
3.54 in.
1 Year