missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BrandTech™ BRAND™ BIO-CERT™ Pipet Tips, 50 to 1000uL

Manufactured from high-purity, oleamide- and diHEMDA-free polypropylene. BrandTech™ BRAND™ BIO-CERT™ are sterile and free of biological contaminants. Thin-walled for high translucency, easy ejection, and low waste. Graduations marked at 250, 500 and 1000μL for quick volume assessment. Available in either standard or ultra low retention types.

$298.89 - $450.80

Specifications

Clean Claims DNase, RNase, ATP, and Endotoxin Free
Color Colorless
Length (Metric) 7cm
Material PP (Polypropylene)
Sterility Sterile
View More Specs

Products 5
Catalog Number Mfr. No. Format Tip Style Price Quantity  
Catalog Number Mfr. No. Format Tip Style Price Quantity  
13-889-163 View Documents BrandTech™
732132
Refill Universal Fit Pack of 960 Each for $438.54
13-889-182 View Documents BrandTech™
732232
Rack Universal Fit Pack of 960 Each for $450.80
13-889-201 View Documents BrandTech™
732332
Rack Low Retention Pack of 960 Each for $298.89
13-889-214 View Documents BrandTech™
732272
Rack Universal Fit Pack of 960 Each for $450.80
13-889-220 View Documents BrandTech™
732372
Rack Low Retention Pack of 960 Each for $411.54
Description

Description

  • High-purity polypropylene is oleamide- and diHEMDA-free
  • Independently certified sterile and free of biological contaminants
  • Thin-walled for high translucency, easy ejection and low waste
  • Graduations at 250, 500 and 1000μL for quick volume check
  • Available in either standard or ultra low retention types
  • Available either in bulk packages or in convenient, ecological rack-and-stack formats
  • Bulk tips are blue colored and racked tips are colorless and placed into blue-colored tip tray

Ultra low retention tips

  • Produced through special, patented physicochemical process
  • Homogeneous, defect-free surface for low surface tension and low sample retention
  • Reduce sample loss and provide substantially higher reproducibility when working with critical media
  • For biological samples that contain proteins, nucleic acids, detergents and other difficult liquids that can increase sample loss
  • High chemical resistance
  • Tips can be autoclaved at 121°C (2 bar) without damaging low retention properties

Specifications

Specifications

DNase, RNase, ATP, and Endotoxin Free
7cm
Sterile
250, 500, 1000μL
Colorless
PP (Polypropylene)
50 to 1000μL
Yes