Please login to your online account to display your discounted pricing
BrandTech™ BRAND™ BIO-CERT™ Pipet Tips, 1 to 50uL
DNA, RNase-, ATP- and endotoxin-free
$257.63 - $334.63
Specifications
Certifications/Compliance DNA, RNase, ATP and endotoxin-free
Material Polypropylene
Product Type Tip
Quantity 960 Pack
Sterility Sterile
View More Specs
Products
Catalog Number Mfr. No. Product Type Packaging Price Quantity    
13-889-160
brandtech™
732126
Tip TipRefill, 10 racks of 96 pieces Pack of 960 Each for $334.63
13-889-179
brandtech™
732226
Tip TipBox, 10 boxes of 96 pieces Pack of 960 Each for $334.63
13-889-198
brandtech™
732326
Tip TipBox, 10 boxes of 96 pieces Pack of 960 Each for $257.63
Description & Specifications
Specifications
Certifications/Compliance DNA, RNase, ATP and endotoxin-free
Material Polypropylene
Product Type Tip
Quantity 960 Pack
Sterility Sterile
Volume (Metric) 1 to 50μL
  • High purity polypropylene is oleamide- and diHEMDA-free
  • Independently certified sterile tips are free from biological contaminants
  • Available in either standard or ultra low retention types
  • Available in convenient, ecological rack format
Ultra low retention tips
  • Produced through special, patented physicochemical process
  • Homogeneous, defect-free surface for low surface tension and low sample retention
  • Reduce sample loss and provide substantially higher reproducibility when working with critical media
  • For biological samples that contain proteins, nucleic acids, detergents and other difficult liquids that can increase sample loss
  • High chemical resistance
  • Tips can be autoclaved at 121°C (2 bar) without damaging low retention properties