Please login to your online account to display your discounted pricing

BrandTech™ BRAND™ BIO-CERT™ nano-cap™ Filter Tips

DNA-, RNase-, ATP- and endotoxin-free

$516.60 - $788.14

Specifications

Certifications/Compliance DNA, RNase, ATP and endotoxin-free
Color Colorless
For Use With (Equipment) Sub-micro Transferpette S
Length (English) 1.45 in.
Length (Metric) 3.7cm
View More Specs
Products
Catalog Number Mfr. No. Product Type Packaging Price Quantity    

13-889-164

 
brandtech™
732622
Filter Tip TipRefill, 10 racks of 96 pieces Pack of 960 Each for $516.60

13-889-183

 
brandtech™
732722
Filter Tip TipBox, 10 boxes of 96 pieces Pack of 960 Each for $788.14

13-889-202

 
brandtech™
732822
Filter Tip TipBox, 10 boxes of 96 pieces Pack of 960 Each for $576.69
Description & Specifications

Specifications

Certifications/Compliance DNA, RNase, ATP and endotoxin-free
Color Colorless
For Use With (Equipment) Sub-micro Transferpette S
Length (English) 1.45 in.
Length (Metric) 3.7cm
Material Polypropylene
Product Type Filter Tip
Quantity 960 Pack
Sterility Sterile
Volume (Metric) 0.1 to 1μL

  • High-purity polypropylene is oleamide- and diHEMDA-free
  • Independently certified sterile and free from biological contaminants
  • Suitable for use with the sub-micro Transferpette S (03-837-317)
  • Features capillary rise that is visible to naked eye even for volumes as small as 0.1μL
  • Available in either standard or ultra low retention types
  • Available in convenient, ecolcogical rack format
  • Tips are colorless and placed into gray-colored tip tray

Ultra low retention tips

  • Produced through special, patented physicochemical process
  • Homogeneous, defect-free surface for low surface tension and low sample retention
  • Reduce sample loss and provide substantially higher reproducibility when working with critical media
  • For biological samples that contain proteins, nucleic acids, detergents and other difficult liquids that can cause sample loss
  • High chemical resistance
  • Tips can be autoclaved at 121°C (2 bar) without damaging low retention or filter properties