Please login to your online account to display your discounted pricing
BrandTech™ BRAND™ BIO-CERT™ Filter Tips, 50 to 1000uL
Constructed of high-purity, oleamide- and diHEMDA-free polypropylene. BrandTech™ BRAND™ BIO-CERT™ Filter Tips, 50 to 1000μL are sterile and free from biological contaminants. Non self-sealing PE filters allow sample recovery with wetted filters. Thin-walled for high translucency, easy ejection, and low waste. Graduations at 250, 500, and 1000μL provide quick volume check. Available in standard or ultra low retention types.
$564.69 - $732.89
Specifications
Certifications/Compliance DNA, RNase, ATP and endotoxin-free
Color Colorless
Length (Metric) 7cm
Material Polypropylene
Sterility Sterile
View More Specs
Products
Catalog Number Mfr. No. Tip Style Packaging Price Quantity    
13-889-189
brandtech™
732734
Standard TipBox, 10 boxes of 96 pieces Pack of 960 Each for $732.89
13-889-208
brandtech™
732834
Ultra Low Retention TipBox, 10 boxes of 96 pieces Pack of 960 Each for $564.69
13-889-170
brandtech™
732634
Standard TipRefill, 10 racks of 96 pieces Pack of 960 Each for $718.49
Description & Specifications
Specifications
Certifications/Compliance DNA, RNase, ATP and endotoxin-free
Color Colorless
Length (Metric) 7cm
Material Polypropylene
Sterility Sterile
Volume (Metric) 50 to 1000μL
Graduations 250, 500, 1000μL
  • High-purity polypropylene is oleamide- and diHEMDA-free
  • Independently certified sterile and free from biological contaminants
  • Non self-sealing PE filters allow sample recovery with wetted filters
  • Thin-walled for high translucency, easy ejection, and low waste.
  • Graduations at 250, 500 and 1000μL for quick volume check.
  • Available in either standard or ultra low retention types
  • Available in convenient, ecological rack format; racked tips are colorless and placed into blue-colored tip tray
Ultra low retention tips
  • Produced through special, patented physicochemical process
  • Homogeneous, defect-free surface for low surface tension and low sample retention
  • Reduce sample loss and provide substantially higher reproducibility when working with critical media
  • For biological samples that contain proteins, nucleic acids, detergents and other difficult liquids that can cause sample loss
  • High chemical resistance
  • Tips can be autoclaved at 121°C (2 bar) without damaging low retention or filter properties