missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BrandTech™ BRAND™ BIO-CERT™ Filter Tips, 5 to 100uL

Manufactured from high-purity, oleamide- and diHEMDA-free polypropylene. BrandTech™ BRAND™ BIO-CERT™ Filter tips, 5 to 100μL allow sample recovery with wetted filters. Sterile tips are free of biological contaminants. Thin-walled for high translucency, easy ejection, and low waste. Graduations marked at 50 and 100μL for quick volume assessment. Available in either standard or ultra low retention types.

$398.03 - $587.26

Specifications

Certifications/Compliance DNA, RNase, ATP and endotoxin-free
Color Colorless
For Use With (Equipment) For pipettes with yellow color code
Length (Metric) 5.3cm
Material Polypropylene
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Price Quantity  
Catalog Number Mfr. No. Packaging Price Quantity  
13-889-168 BrandTech™
732630
Rack, TipRefill 10 x 96 Pack of 960 Each for $398.03
13-889-187 BrandTech™
732730
Rack, TipBox 10 x 96 Pack of 960 Each for $587.26
13-889-206 BrandTech™
732830
Rack, TipBox 10 x 96 Pack of 960 Each for $447.17
Specifications

Description

  • Non-sealing PE filters allow sample recovery with wetted filters
  • High-purity polypropylene is oleamide- and diHEMDA-free
  • Independently certified sterile tips are DNA-, RNase, ATP- and endotoxin-free
  • Thin-walled for high translucency, easy ejection and low waste
  • Graduations at 50 and 100μL for quick volume check
  • Available in either standard or ultra low retention types
  • Available in convenient, ecological rack format, racked tips are colorless and placed into green-colored tip tray

Ultra low retention tips

  • Produced through special, patented physicochemical process
  • Homogeneous, defect-free surface for low surface tension and low sample retention
  • Reduce sample loss and provide substantially higher reproducibility when working with critical media
  • For biological samples that contain protein, nucleic acid, detergents or other difficult liquids that can result in sample loss
  • High chemical resistance
  • Tips can be autoclaved at 121°C (2 bar) without damaging low retention and filter properties

Description & Specifications

Specifications

DNA, RNase, ATP and endotoxin-free
For pipettes with yellow color code
Polypropylene
5 to 100μL
Yes
Colorless
5.3cm
Sterile
50, 100μL