Please login to your online account to display your discounted pricing

BrandTech™ BRAND™ BIO-CERT™ Filter Tips, 1 to 20uL

DNA-, RNase-, ATP- and endotoxin-free

$459.46 - $587.26

Specifications

Certifications/Compliance DNA, RNase, ATP and endotoxin-free
Color Colorless
Material Polypropylene
Product Type Filter Tip
Quantity 960 Pack
View More Specs
Products
Catalog Number Mfr. No. Product Type Packaging Price Quantity    

13-889-166

 
brandtech™
732626
Filter Tip TipRefill, 10 racks of 96 pieces Pack of 960 Each for $474.20

13-889-185

 
brandtech™
732726
Filter Tip TipBox, 10 boxes of 96 pieces Pack of 960 Each for $587.26

13-889-204

 
brandtech™
732826
Filter Tip TipBox, 10 boxes of 96 pieces Pack of 960 Each for $459.46
Description & Specifications

Specifications

Certifications/Compliance DNA, RNase, ATP and endotoxin-free
Color Colorless
Material Polypropylene
Product Type Filter Tip
Quantity 960 Pack
Sterility Sterile
Volume (Metric) 1 to 20μL
For Use With (Application) High translucency, easy ejection and low waste, quick volume check
Graduations 1, 20μL

  • High-purity polypropylene is oleamide- and diHEMDA-free
  • Independently certified sterile and free of biological contaiminants
  • Available in either standard or ultra low retention types
  • Available in convenient, ecological rack format

Ultra low retention tips

  • Produced through special, patented physicochemical process
  • Homogeneous, defect-free surface for low surface tension and low sample retention
  • Reduce sample loss and provide substantially higher reproducibility when working with critical media
  • For biological samples that contain protein, nucleic acids, detergents and other difficult liquids that can result in sample loss
  • High chemical resistance
  • Tips can be autoclaved at 121°C (2 bar) without damaging low retention or filter properties