Please login to your online account to display your discounted pricing
BrandTech™ BRAND™ BIO-CERT™ Filter Tips, 0.5 to 10uL
Manufactured from high-purity, oleamide- and diHEMDA-free polypropylene. BrandTech™ BRAND™ BIO-CERT™ Filter Tips, 0.5 to 10μL are sterile and free of biological contaminants. Non self-sealing PE filters allow sample recovery with wetted filters. Slender design allows pipetting without touching microtube and microplate walls. Graduations provided at 2 and 10μL for quick volume check.
$449.77 - $574.86
Specifications
Certifications/Compliance DNA, RNase, ATP and endotoxin-free
Color Colorless
Length (Metric) 4.6cm
Material Polypropylene
Sterility Sterile
View More Specs
Products
Catalog Number Mfr. No. Tip Style Packaging Price Quantity    
13-889-184
brandtech scientific
732724
Standard TipBox, 10 boxes of 96 pieces Pack of 960 Each for $574.86
13-889-203
brandtech scientific
732824
Ultra Low Retention, Nano-cap TipBox, 10 boxes of 96 pieces Pack of 960 Each for $449.77
13-889-165
brandtech scientific
732624
Standard TipRefill, 10 racks of 96 pieces Pack of 960 Each for $464.17
Description & Specifications
Specifications
Certifications/Compliance DNA, RNase, ATP and endotoxin-free
Color Colorless
Length (Metric) 4.6cm
Material Polypropylene
Sterility Sterile
Graduations 2, 10μL
  • High-purity polypropylene is oleamide- and diHEMDA-free
  • Independently certified sterile and free of biological contaminants
  • Non self-sealing PE filters allow sample recovery with wetted filters
  • Slender design
  • Thin-walled for high translucency, easy ejection, and low waste
  • Graduations at 2 and 10μL for quick volume check
  • Available in either standard or ultra low retention types
  • Available in convenient, ecological rack format, racked tips are colorless and placed into gray-colored tip tray
Ultra low retention tips
  • Produced through special, patented physicochemical process
  • Homogeneous, defect-free surface for low surface tension and low sample retention
  • Reduce sample loss and provide substantially higher reproducibility when working with critical media
  • For biological samples that contain proteins, nucleic acids, detergents and other difficult liquids that can cause sample retention
  • High chemical resistance
  • Tips can be autoclaved at 121°C (2 bar) without damaging low retention or filter properties