Please login to your online account to display your discounted pricing

BrandTech™ BRAND™ 96 Well PCR Plates

Manufactured from high-quality virgin plastics for excellent, reproducible results in most PCR applications

$541.24 - $668.88

Specifications

Autoclavable Yes
Certifications/Compliance RNase, DNase-free
For Use With (Application) PCR
Color Clear
Material Polypropylene
View More Specs
Products
Catalog Number Mfr. No. Skirt Style Price Quantity    

13-882-74

 
brandtech™
781350
Non-skirt Pack of 50 Each for $541.24

13-882-75

 
brandtech™
781400
Half Pack of 50 Each for $668.88
Description & Specifications

Specifications

Autoclavable Yes
Certifications/Compliance RNase, DNase-free
For Use With (Application) PCR
Color Clear
Material Polypropylene

  • Manufactured without lubricants or chemical additives that can contaminate reagents and samples
  • Autoclavable and RNase-, DNase-, and DNA-free
  • Thin, uniform walls ensure consistent, precise thermal transfer and excellent results
  • Compatible with most major brands of thermal cyclers
  • 96 wells