BrandTech™ BRAND™ BLAUBRAND™ Type 2 Glass Graduated Pipets, Class A, USP, Certified
In DIN EN ISO 835 standard, waiting time has been reduced from 15 to 5 seconds
Manufacturer:  BrandTech™ BRAND™ 27513
 View more versions of this product
Catalog No. 13-690-126
Description
Description
  • Made of AR-GLAS
  • Only requires setting meniscus one time
  • Contrast blue enamel ensures optimum combination on resistance and readability
  • Features full ring marks and major graduations
  • Flexible screen stencils for volume markings to match calibration marks for enhanced accuracy at all intermediate volumes
  • Optimized flow geometry for reliable liquid dispensing from pipet
  • Fire-polished edges for durability
  • Available in all ISO sizes from 1 to 10mL
  • Supplied with batch number and an accompanying USP batch certificate in original packaging
Specifications
Specifications
USP
Yes
Type 2 graduated pipet
AR-GLAS™
A
0.1mL
10mL
Documents