Please login to your online account to display your discounted pricing

BrandTech™ Analog Dispensette™ Organic Bottle-Top Dispensers

Ideal for HPLC and combinatorial chemistry labs

$1,221.77 - $1,221.77

Specifications

Autoclavable Yes
Certifications/Compliance ISO
Density Up to 2.2g/cm3
Product Type Dispensette™ Organic Bottletop Dispenser
Display Analog
View More Specs
Includes:  Discharge tube, valve, telescopic filling tube, selection of bottle-top thread adapters, manual, and calibration certificate
Products
Catalog Number Mfr. No. Accuracy Increments Volume Range Price Quantity    

13-688-270

  Accessories
brandtech™
4731131
≤0.5% ±25μL 0.1mL 0.5 to 5mL Each for $1,221.77
Description & Specifications

Specifications

Autoclavable Yes
Certifications/Compliance ISO
Density Up to 2.2g/cm3
Product Type Dispensette™ Organic Bottletop Dispenser
Display Analog
Max. Pressure Up to 600mbar
Operating Temperature (English) 59° to 104°F
Operating Temperature (Metric) 15° to 40°C
Viscosity Rating 500mm2/sec.
Warranty 1 Year

For dispensing organic solvents including chlorinated and fluorinated hydrocarbons, concentrated acids (e.g., HNO3), trifluoroacetic acid (TFA) tetrahydrofuran (THF), methanol, acetonitrile, methylene choride, DMF, and DMSO.

  • Mounts directly on most solvent and reagent bottles for fast, convenient dispensing
  • Reinforced discharge tube with threaded safety cap
  • Safety discharge system to reduce the chance of inadvertent dispensing
  • Telescoping filling tube and polypropylene thread adapters (ETFE adapters available separately)
  • Easily disassemble for cleaning and autoclaving
  • Analog and adjustable Optional SafetyPrime™ valve reduces spurting and reagent waste while priming

  • Flowpath consists of borosilicate glass, ceramic, tantalum, PTFE, ETFE, FEP, and PFA
  • Discharge tube safety screw cap is polypropylene

Must not be used for hydrofluoric acid (HF), carbon disulfide, alkaline solutions, hydrochloride salts, or liquids incompatible with the listed flowpath materials.