Invitrogen™ BRAF Recombinant Human Protein DFS Item
BRAF is involved in the transduction of mitogenic signals from the cell membrane to the nucleus.
Manufacturer:  Invitrogen™ LSPV4172
 View more versions of this product
Catalog No. PV4172
Description
Description
BRAF is activated by somatic point mutation in human cancer, and is thought to play a role in the postsynaptic responses of hippocampal neuron.
Specifications
Specifications
NP_004324.2
Frozen
113.6kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
RAF Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
BRAF, BRAF1
Baculovirus
Human
GST Tag
Documents