Please login to your online account to display your discounted pricing

Alfa Aesar™ Bradykinin (2-7)

For cellular and molecular biology applications

Specifications

Product Type Bradykinin (2-7)
Molecular Weight 600.66
Storage Requirements -30° to -10°C
Sequence Pro-Pro-Gly-Phe-Ser-Pro
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66151LB0

 
alfa aesar™
J66151LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66151MB

 
alfa aesar™
J66151MB
25mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Bradykinin (2-7)
Molecular Weight 600.66
Storage Requirements -30° to -10°C
Sequence Pro-Pro-Gly-Phe-Ser-Pro
Format Solid
Regulatory Status RUO
Purification Method HPLC