Please login to your online account to display your discounted pricing

Alfa Aesar™ Bradykinin (1-6)


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66937LB0

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ66937LB0

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Quantity 5mg
Format Solid
Sequence Arg-Pro-Pro-Gly-Phe-Ser
Regulatory Status RUO
Storage Requirements -30° to -10°C
Product Type Bradykinin (1-6)
Molecular Weight 659.73
Purification Method HPLC