Please login to your online account to display your discounted pricing

Alfa Aesar™ Bradykinin (1-5)

For cellular and molecular biology applications

Specifications

Product Type Bradykinin (1-5)
Molecular Weight 572.67
Storage Requirements -30° to -10°C
Sequence H-Arg-Pro-Pro-Gly-Phe-OH
For Use With (Application) Biologically active fragment of bradykinin
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ63233LB0

 
alfa aesar™
J63233LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Bradykinin (1-5)
Molecular Weight 572.67
Storage Requirements -30° to -10°C
Sequence H-Arg-Pro-Pro-Gly-Phe-OH
For Use With (Application) Biologically active fragment of bradykinin
Format Powder
Regulatory Status RUO
Purification Method HPLC